Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(R)-(-)-1-[(S)-2-(Di-1-naphthylphosphino)ferrocenyl]ethyldi-3,5-xylylphosphine ≥97%

Warning

26-1175-100MG 26-1175-500MG
101442-180EA 157.11 USD InStock
101442-180 101442-182
(R)-(-)-1-[(S)-2-(Di-1-naphthylphosphino)ferrocenyl]ethyldi-3,5-xylylphosphine ≥97%
(R)-(-)-1-[(S)-2-(Di-1-naphthylphosphino)ferrocenyl]ethyldi-3,5-xylylphosphine
JOSIPHOS, Phosphine


Formula: C₄₈H₄₄FeP₂
MW: 738.660 g/mol
MDL Number: MFCD08561163
CAS Number: 851308-40-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty