Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ferrocenecarboxylic acid ≥97%

Warning

26-1900-1G 26-1900-25G 26-1900-5G
100197-468EA 66.52 USD InStock
100197-468 100207-246 100203-760
Ferrocenecarboxylic acid ≥97%
Ferrocenecarboxylic acid


Formula: C₁₁H₁₀FeO₂
Melting Pt: 210 °C (dec.)
MDL Number: MFCD00001430
CAS Number: 1271-42-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty