Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Bis(6,6,7,7,8,8,8-heptafluoro-2,2-dimethyl-3,5-octanedionate)copper(II)


29-8400-1G 29-8400-5G
100200-920EA 169.85 USD InStock
100200-920 100197-108
Bis(6,6,7,7,8,8,8-heptafluoro-2,2-dimethyl-3,5-octanedionate)copper(II)
Bis(6,6,7,7,8,8,8-heptafluoro-2,2-dimethyl-3,5-octanedionate)copper(II)


Formula: Cu(C₁₀H₁₀F₇O₂)₂
MW: 653.9200 g/mol
Boiling Pt: 280 °C/ 100 °C (0.1 mmHg) (dec./ subl.)
Melting Pt: 68…69 °C
MDL Number: MFCD00156516
CAS Number: 80289-21-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty