Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Bis(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')zinc(II) ≥99%


30-0500-25G 30-0500-5G 30-0500-1G
100205-926EA 1282.34 USD InStock
100205-926 100204-034 100200-454
Bis(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')zinc(II) ≥99%
Bis(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')zinc(II)
Metal Beta-diketonates, Metal TMHD, Volatile Precursors for CVD


Formula: C₂₂H₃₈O₄Zn
MW: 431.92 g/mol
Boiling Pt: 250 °C (dec.)
Melting Pt: 144 °C
MDL Number: MFCD00192578
CAS Number: 14363-14-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty