Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Bis(isopropylcyclopentadienyl)zirconium dichloride ≥98%

Warning

40-1050-1G 40-1050-5G
100200-946EA 174.09 USD InStock
100200-946 100201-214
Bis(isopropylcyclopentadienyl)zirconium dichloride ≥98%
Bis(isopropylcyclopentadienyl)zirconium dichloride
Metallocenes, Derivatives & Cp Precursors


Formula: C₁₆H₂₂Cl₂Zr
MDL Number: MFCD00269855
CAS Number: 58628-40-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty