Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

rac-Dimethylsilylbis(1-indenyl)zirconium dichloride ≥97%

Warning

40-1142-500MG 40-1142-100MG
100200-954EA 590.22 USD InStock
100200-954 100200-952
rac-Dimethylsilylbis(1-indenyl)zirconium dichloride ≥97%
rac-Dimethylsilylbis(1-indenyl)zirconium dichloride
Metallocenes, Derivatives & Cp Precursors


MDL Number: MFCD00672108
CAS Number: 121009-93-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty