Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,6-Diisopropylphenylimidoneophylidene molybdenum(VI) bis(t-butoxide)

Warning

42-1200-500MG 42-1200-100MG
100210-646EA 839.32 USD InStock
100210-646 100209-930
2,6-Diisopropylphenylimidoneophylidene molybdenum(VI) bis(t-butoxide)
2,6-Diisopropylphenylimidoneophylidene molybdenum(VI) bis(t-butoxide)
Alkoxide, Schrock's Catalyst


MDL Number: MFCD00142847
CAS Number: 126949-65-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty