Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Dichlorodicarbonylbis(triphenylphosphine)ruthenium(II) ≥98%

Warning

44-0400-1G 44-0400-250MG
100200-684EA 386.4 USD InStock
100200-684 100205-126
Dichlorodicarbonylbis(triphenylphosphine)ruthenium(II) ≥98%
Dichlorodicarbonylbis(triphenylphosphine)ruthenium(II)


Formula: C₃₈H₃₄Cl₂O₂P₂Ru
MDL Number: MFCD00009594
CAS Number: 14564-35-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty