Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Dichloro(p-cymene)tricyclohexylphosphineruthenium(II) ≥97%

Warning

44-0430-250MG 44-0430-1G
100196-220EA 117.48 USD InStock
100196-220 100206-650
Dichloro(p-cymene)tricyclohexylphosphineruthenium(II) ≥97%
Dichloro(p-cymene)tricyclohexylphosphineruthenium(II)


MDL Number: MFCD06798308
CAS Number: 145381-23-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty