Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ruthenium(VIII) oxide 0,5% in aqueous solution stabilised

Danger

44-2000-50ML 44-2000-10ML
100201-654EA 901.6 USD InStock
100201-654 100201-652
Ruthenium(VIII) oxide 0,5% in aqueous solution stabilised
Ruthenium(VIII) oxide


Formula: O₄Ru
MDL Number: MFCD00074857
CAS Number: 20427-56-9
UN: 3139
ADR: 5.1,II
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty