Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Tetrabutylammonium perruthenate ≥97%

Danger

44-7760-2G 44-7760-500MG
100203-786EA 567.57 USD InStock
100203-786 100196-234
Tetrabutylammonium perruthenate ≥97%
Tetrabutylammonium perruthenate


Melting Pt: 157 °C (dec.)
MDL Number: MFCD02684567
CAS Number: 96317-72-5
UN: 1479
ADR: 5.1,II
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty