Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Tris(acetonitrile)pentamethylcyclopentadienylruthenium(II) hexafluorophosphate ≥98%

Warning

44-7880-250MG 44-7880-1G
100211-104EA 237.78 USD InStock
100211-104 100199-918
Tris(acetonitrile)pentamethylcyclopentadienylruthenium(II) hexafluorophosphate ≥98%
Tris(acetonitrile)pentamethylcyclopentadienylruthenium(II) hexafluorophosphate
Metallocenes, Derivatives & Cp Precursors


Formula: C₁₆H₂₄N₃PRuF₆
MDL Number: MFCD04117715
CAS Number: 99604-67-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty