Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(+)-1,2-Bis((2S,5S)-2,5-dimethylphospholano)benzene(1,5-cyclooctadiene)rhodium(I)trifluoromethanesulfonate ≥98%

Warning

45-0161-2G 45-0161-500MG 45-0161-100MG
100197-146EA 2516.55 USD InStock
100197-146 100208-658 100208-656
(+)-1,2-Bis((2S,5S)-2,5-dimethylphospholano)benzene(1,5-cyclooctadiene)rhodium(I)trifluoromethanesulfonate ≥98%
(+)-1,2-Bis((2S,5S)-2,5-dimethylphospholano)benzene(1,5-cyclooctadiene)rhodium(I)trifluoromethanesulfonate
BPE, DUPHOS


Formula: C₂₇H₄₀F₃O₃P₂RhS
MW: 666.5179 g/mol
MDL Number: MFCD00269863
CAS Number: 136705-75-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty