Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Norbornadiene rhodium(I) chloride dimer ≥99%

Warning

45-0500-1G 45-0500-250MG
100196-950EA 1021.91 USD InStock
100196-950 100200-982
Norbornadiene rhodium(I) chloride dimer ≥99%
Norbornadiene rhodium(I) chloride dimer


Formula: C₁₄H₁₆Cl₂Rh₂
MDL Number: MFCD00198060
CAS Number: 12257-42-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty