Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Rhodium 5% on carbon

Danger

45-1860-25G 45-1860-1G 45-1860-5G
100197-528EA 3122.34 USD InStock
100197-528 100201-664 100201-666
Rhodium 5% on carbon
Rhodium


Formula: Rh
MW: 102.9
Melting Pt: 1960
Density: 12.41
MDL Number: MFCD00011201
CAS Number: 7440-16-6
UN: 3178
ADR: 4.1 II
Merck Index: 12,08353
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty