Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Bis(2-methylallyl)palladium chloride dimer ≥98%

Warning

46-0240-500MG 46-0240-2G
100206-702EA 150.03 USD InStock
100206-702 100210-744
Bis(2-methylallyl)palladium chloride dimer ≥98%
Bis(2-methylallyl)palladium chloride dimer


Melting Pt: 168…170 °C
MDL Number: MFCD02684573
CAS Number: 12081-18-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty