Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Dichloro(di-mu-chloro)bis[1,3-bis(2,6-di-i-propylphenyl)imidazol-2-ylidene]dipalladium(II) ≥97%

Warning

46-0860-1G 46-0860-250MG
101419-070EA 362.34 USD InStock
101419-070 101419-068
Dichloro(di-mu-chloro)bis[1,3-bis(2,6-di-i-propylphenyl)imidazol-2-ylidene]dipalladium(II) ≥97%
Dichloro(di-mu-chloro)bis[1,3-bis(2,6-di-i-propylphenyl)imidazol-2-ylidene]dipalladium(II)
NHC


Formula: C₅₄H₇₂Cl₄N₄Pd₂
MDL Number: MFCD09039100
CAS Number: 444910-17-2
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty