Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Palladium 10% on activated carbon wetted with 50-70% water, powder Pearlman, Evonik Noblyst® P1070

Warning

46-1706-10G 46-1706-50G
100198-828EA 437.35 USD InStock
100198-828 100198-830
Palladium 10% on activated carbon wetted with 50-70% water, powder Pearlman, Evonik Noblyst® P1070
Palladium


Formula: Pd
MW: 106.420 g/mol
MDL Number: MFCD00011167
CAS Number: 7440-05-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty