Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Palladium 5% on silica, dried powder, reduced (Escat™ 1351)


46-2090-25G 46-2090-5G
100206-722EA 1167.69 USD InStock
100206-722 100206-724
Palladium 5% on silica, dried powder, reduced (Escat™ 1351)
Palladium
Escat


Formula: Pd
MDL Number: MFCD00011167
CAS Number: 7440-05-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty