Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Tris(dibenzylideneacetone)dipalladium(0), complex with chloroform


46-3010-2G 46-3010-500MG
100199-950EA 267.51 USD InStock
100199-950 100211-124
Tris(dibenzylideneacetone)dipalladium(0), complex with chloroform
Tris(dibenzylideneacetone)dipalladium(0), complex with chloroform


Formula: C₅₁H₄₂O₃Pd₂·CHCl₃
Melting Pt: 131…135 °C
MDL Number: MFCD00075479
CAS Number: 52522-40-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty