Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Tin(II) trifluoromethanesulphonate ≥99%

Warning

50-2750-10G 50-2750-2G
100206-748EA 387.82 USD InStock
100206-748 100206-750
Tin(II) trifluoromethanesulphonate ≥99%
Tin(II) trifluoromethanesulphonate
Metal Triflate


Formula: C₂F₆O₆S₂Sn
MW: 416.85 g/mol
Melting Pt: Dec.
MDL Number: MFCD00191251
CAS Number: 62086-04-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty