Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Antimony(IV) oxide ≥99.99% (Sb basis), PURATREM

Danger

51-5110-5G 51-5110-25G
100199-960EA 69.35 USD InStock
100199-960 100208-034
Antimony(IV) oxide ≥99.99% (Sb basis), PURATREM
Antimony(IV) oxide


Density: 5.82
MDL Number: MFCD00011215
CAS Number: 1332-81-6
UN: 1549
ADR: 6.1,III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty