Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Cesium 2-ethylhexoxide 0.8-1.0M in octane/toluene

Danger

55-2000-25G 55-2000-5G
100203-230EA 533.6 USD InStock
100203-230 100203-232
Cesium 2-ethylhexoxide 0.8-1.0M in octane/toluene
Cesium 2-ethylhexoxide
Alkoxide


Formula: C₈H₁₇CsO
Flash Pt: 60 °F
MDL Number: MFCD00143940
CAS Number: 259743-14-1
UN: 1993
ADR: 3,II
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty