Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Bis(6,6,7,7,8,8,8-heptafluoro-2,2-dimethyl-3,5-octanedionate)barium

Warning

56-8400-25G 56-8400-1G 56-8400-5G
100207-414EA 1957.48 USD InStock
100207-414 100210-018 100210-020
Bis(6,6,7,7,8,8,8-heptafluoro-2,2-dimethyl-3,5-octanedionate)barium
Bis(6,6,7,7,8,8,8-heptafluoro-2,2-dimethyl-3,5-octanedionate)barium
Metal Beta-diketonates, Volatile Precursors for CVD


Boiling Pt: 280…300 °C/ 210 °C (0,2 mmHg) (dec./ subl.)
Melting Pt: 194…198 °C
MDL Number: MFCD00070455
CAS Number: 36885-31-1
UN: 1564
ADR: 6.1,III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty