Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)lanthanum(III) tetraglyme adduct ≥99.9% (La basis)


57-1100-25G 57-1100-5G 57-1100-1G
100211-140EA 2966.65 USD InStock
100211-140 100211-142 100211-138
Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)lanthanum(III) tetraglyme adduct ≥99.9% (La basis)
Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)lanthanum(III) tetraglyme adduct
Metal Beta-diketonates, Metal TMHD, Volatile Precursors for CVD


Boiling Pt: 125 °C (0.1 mmHg)
Melting Pt: 59 °C
MDL Number: MFCD01073801
CAS Number: 151139-14-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty