Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Tetrabenzylhafnium ≥97%

Danger

72-7700-500MG 72-7700-2G
100206-792EA 179.75 USD InStock
100206-792 100206-794
Tetrabenzylhafnium ≥97%
Tetrabenzylhafnium


MDL Number: MFCD03427339
CAS Number: 31406-67-4
UN: 3178
ADR: 4.1,II
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty