Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyltrioxorhenium(VII) ≥98%


75-2375-5G 75-2375-250MG 75-2375-1G
100208-954EA 1193.17 USD InStock
100208-954 100211-162 100208-952
Methyltrioxorhenium(VII) ≥98%
Methyltrioxorhenium(VII)


Formula: CH₃O₃Re
MW: 249.24 g/mol
Boiling Pt: 65 °C (0.001 mmHg) (subl.)
Melting Pt: 111 °C
MDL Number: MFCD00192332
CAS Number: 70197-13-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty