Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(Pentamethylcyclopentadienyl)iridium(III) dichloride dimer ≥98%

Warning

77-1060-2G 77-1060-500MG
100198-254EA 1246.95 USD InStock
100198-254 100199-986
(Pentamethylcyclopentadienyl)iridium(III) dichloride dimer ≥98%
(Pentamethylcyclopentadienyl)iridium(III) dichloride dimer
CATHy Catalyst, Metallocenes, Derivatives & Cp Precursors


Formula: C₂₀H₃₀Cl₄Ir₂
MW: 796.69 g/mol
MDL Number: MFCD00075435
CAS Number: 12354-84-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty