Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(Trimethyl)cyclopentadienylplatinum(IV) ≥99%


78-1300-2G 78-1300-500MG
101324-828EA 1193.17 USD InStock
101324-828 101324-826
(Trimethyl)cyclopentadienylplatinum(IV) ≥99%
(Trimethyl)cyclopentadienylplatinum(IV)
Metallocenes, Derivatives & Cp Precursors, Volatile Precursors for CVD


Melting Pt: 104…106 °C
MDL Number: MFCD07369041
CAS Number: 1271-07-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty