Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Potassium bis(oxalato)platinate(II) dihydrate ≥99%

Warning

78-1935-5G 78-1935-1G
100199-260EA 1612.12 USD InStock
100199-260 100203-312
Potassium bis(oxalato)platinate(II) dihydrate ≥99%
Potassium bis(oxalato)platinate(II) dihydrate


Formula: C₄H₄K₂O₁₀Pt
MW: 485.343 g/mol
MDL Number: MFCD00167153
CAS Number: 14244-64-5
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty