Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

cis-Tetrachlorodiammine platinum(IV) ≥99%

Danger

78-2024-5G 78-2024-250MG 78-2024-1G
100211-178EA 1337.54 USD InStock
100211-178 100197-618 100211-176
cis-Tetrachlorodiammine platinum(IV) ≥99%
cis-Tetrachlorodiammine platinum(IV)


Formula: CL₄H₆N₂Pt
MDL Number: MFCD00050045
CAS Number: 16893-05-3
UN: 2811
ADR: 6.1,III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty