Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

trans-Tetrachlorodiammine platinum(IV) ≥98%

Danger

78-2025-250MG 78-2025-1G
100200-720EA 110.4 USD InStock
100200-720 100198-900
trans-Tetrachlorodiammine platinum(IV) ≥98%
trans-Tetrachlorodiammine platinum(IV)


Formula: Cl₄H₄N₂Pt
MDL Number: MFCD00050045
CAS Number: 16893-06-4
UN: 2811
ADR: 6.1,III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty