Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Thallium(I) 2-ethylhexanoate (56 - 59% Tl), Superconductor Grade

Danger

81-2400-100G 81-2400-25G
100201-040EA 1229.97 USD InStock
100201-040 100198-264
Thallium(I) 2-ethylhexanoate (56 - 59% Tl), Superconductor Grade
Thallium (I) 2-ethylhexanoate


Formula: C₈H₁₅O₂Tl
MW: 347.587 g/mol
MDL Number: MFCD00070466
CAS Number: 210578-56-6
UN: 1707
ADR: 6.1 II
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty