Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Bismuth(III) 2-ethylhexanoate 70-75% in xylene (~24% Bi), PURATREM

Warning

83-2401-100G 83-2401-25G
100196-978EA 1337.54 USD InStock
100196-978 100201-046
Bismuth(III) 2-ethylhexanoate 70-75% in xylene (~24% Bi), PURATREM
Bismuth (III) 2-ethylhexanoate


Formula: C₂₄H₄₅BiO₆
MW: 638.598 g/mol
MDL Number: MFCD00043803
CAS Number: 67874-71-9
UN: 1993
ADR: 3 III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty