Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Lithium diethylamide ≥95%

Danger

93-0317-5G 93-0317-25G
100205-234EA 89.17 USD InStock
100205-234 100199-464
Lithium diethylamide ≥95%
Lithium diethylamide


MDL Number: MFCD00040489
CAS Number: 816-43-3
UN: 2846
ADR: 4.2 I
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty