Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Boron trifluoride acetic acid complex (1:2) ≥97%

Danger

93-0517-500G 93-0517-100G
100205-258EA 239.2 USD InStock
100205-258 100198-938
Boron trifluoride acetic acid complex (1:2) ≥97%
Boron trifluoride acetic acid complex (1:2)


Formula: C₄H₈BF₃O₄
MW: 187.91 g/mol
Melting Pt: –47 °C
Density: 1.353
Flash Pt: 181.4 °F
MDL Number: MFCD00036359
CAS Number: 373-61-5
UN: 1742
ADR: 8 II
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty