Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Borane dimethylamine complex ≥97%


93-0554-10G 93-0554-50G
100207-760EA 45.29 USD InStock
100207-760 100202-524
Borane dimethylamine complex ≥97%
Borane dimethylamine complex


Formula: C₂H₁₀BN
MDL Number: MFCD00051068
CAS Number: 74-94-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty