Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Sodium-tert-butanolate ≥98%

Danger

93-1022-100G 93-1022-500G
100208-998EA 80.68 USD InStock
100208-998 100202-526
Sodium-tert-butanolate ≥98%
Sodium-tert-butanolate
Alkoxide


Formula: C₄H₉NaO
MW: 96.1 g/mol
MDL Number: MFCD00040575
CAS Number: 865-48-5
UN: 3206
ADR: 4.2 II
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty