Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Magnesium hexafluoroacetylacetonate dihydrate ≥97%

Warning

93-1227-5G 93-1227-1G
100210-166EA 236.37 USD InStock
100210-166 100196-480
Magnesium hexafluoroacetylacetonate dihydrate ≥97%
Magnesium hexafluoroacetylacetonate dihydrate


Formula: C₁₀H₂F₁₂MgO₄·2H₂O
MDL Number: MFCD05861444
CAS Number: 120156-45-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty