Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Aluminium tri-sec-butanolate ≥98%

Danger

93-1308-100G 93-1308-450G
100204-168EA 53.78 USD InStock
100204-168 100210-178
Aluminium tri-sec-butanolate ≥98%
Aluminium tri-sec-butanolate
Alkoxide, Metal Alkoxides, Volatile Precursors for CVD


Formula: C₁₂H₂₇AlO₃
MW: 246.33 g/mol
Boiling Pt: 200…206 °C (30 mmHg)
Density: 0.9671
Flash Pt: 82 °F
MDL Number: MFCD00009327
CAS Number: 2269-22-9
UN: 2924
ADR: 3 III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty