Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Dimethoxymethylphenylsilane ≥97%


93-1416-100G 93-1416-25G
100201-912EA 338.28 USD InStock
100201-912 100197-700
Dimethoxymethylphenylsilane ≥97%
Dimethoxymethylphenylsilane


Formula: C₉H₁₄O₂Si
MW: 182.29 g/mol
Boiling Pt: 199…200 °C
Density: 0.993
Flash Pt: 177 °F
MDL Number: MFCD00008343
CAS Number: 3027-21-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty