Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Tris(trimethylsilyl)amine ≥98%

Warning

93-1469-25G 93-1469-5G
100206-010EA 352.43 USD InStock
100206-010 100201-920
Tris(trimethylsilyl)amine ≥98%
Tris(trimethylsilyl)amine


Formula: C₉H₂₇NSi₃
Boiling Pt: 76 °C (12 mmHg)
Melting Pt: 70…71 °C
Density: 0,8635
MDL Number: MFCD00047990
CAS Number: 1586-73-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty