Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Polyphosphoric acid (83% P₂O₅)

Danger

93-1552-1000G 93-1552-250G
100209-026EA 203.82 USD InStock
100209-026 100210-214
Polyphosphoric acid (83% P₂O₅)
Polyphosphoric acid


Formula: H(PO₃H)nOH
MDL Number: MFCD00084480
CAS Number: 8017-16-1
UN: 3264
ADR: 8 II
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty