Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Potassium metaphosphate ≥98%


93-1941-500G 93-1941-2000G
100196-528EA 229.29 USD InStock
100196-528 100205-386
Potassium metaphosphate ≥98%
Potassium metaphosphate


Melting Pt: 807 °C
Density: 2,393
MDL Number: MFCD00049681
CAS Number: 7790-53-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty