Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Calcium molybdate ≥99.9% (Ca basis)


93-2026-100G 93-2026-25G
100204-218EA 107.57 USD InStock
100204-218 100210-242
Calcium molybdate ≥99.9% (Ca basis)
Calcium molybdate


Formula: CaMoO₄
MDL Number: MFCD00015976
CAS Number: 7789-82-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty