Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Copper(II) formate hydrate

Warning

93-2925-50G 93-2925-10G
100200-116EA 447.26 USD InStock
100200-116 100207-014
Copper(II) formate hydrate
Copper(II) formate hydrate


Formula: C₂H₂CuO₄·xH₂O
MW: 153.55 g/mol (anhy)
Density: 1.831
MDL Number: MFCD00150509
CAS Number: 544-19-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty