Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Strontium acetate, Reagent Grade


93-3801-100G 93-3801-500G
100200-132EA 69.35 USD InStock
100200-132 100200-134
Strontium acetate, Reagent Grade
Strontium acetate


Formula: C₄H₆O₄Sr
Density: 2.099
MDL Number: MFCD00036392
CAS Number: 543-94-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty