Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Yttrium(III) perchlorate 50% in aqueous solution ≥99.9% (Y basis)


93-3913-50G 93-3913-10G
100200-756EA 532.18 USD InStock
100200-756 100209-110
Yttrium(III) perchlorate 50% in aqueous solution ≥99.9% (Y basis)
Yttrium(III) perchlorate


Formula: Y(ClO₄)₃
MW: 387.26 g/mol
MDL Number: MFCD00150430
CAS Number: 14017-56-2
UN: 3211
ADR: 5.1 II
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty