Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Silver phosphate ≥99%


93-4725-10G 93-4725-50G
100196-678EA 84.92 USD InStock
100196-678 100196-680
Silver phosphate ≥99%
Silver phosphate


Formula: Ag₃O₄P
MW: 418.58 g/mol
Melting Pt: 849 °C
Density: 6.37
MDL Number: MFCD00014148
CAS Number: 7784-09-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty