Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Indium (III) nitrate pentahydrate ≥99.999% (In basis), PURATREM

Danger

93-4923-50G 93-4923-10G
100200-158EA 526.52 USD InStock
100200-158 100203-496
Indium (III) nitrate pentahydrate ≥99.999% (In basis), PURATREM
Indium (III) nitrate pentahydrate


MDL Number: MFCD00149742
CAS Number: 13465-14-0
UN: 1477
ADR: 5.1,II
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty